COM448628633 Combi-Blocks, Inc.

(Z)-4-(3,4-Dimethoxyphenylamino)-4-oxobut-2-enoic acid

  • MDL number MFCD01185849
  • Cas Number : 468747-44-6
  • Molecular Weight : 251.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9626

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9626
Availability details for
Quantity
  • Availability
  • Add to Cart