COM448628645 Combi-Blocks, Inc.

[1-(2-Amino-ethyl)-cyclohexylmethyl]-carbamic acid tert-butyl ester

  • MDL number MFCD08272985
  • Cas Number : 886362-17-0
  • Molecular Weight : 256.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9641

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9641
Availability details for
Quantity
  • Availability
  • Add to Cart