COM448628682 Combi-Blocks, Inc.

[4-(tert-Butyl-dimethyl-silanyloxymethyl)phenyl]-acetic acid

  • MDL number MFCD06796530
  • Cas Number : 886363-54-8
  • Molecular Weight : 280.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9685

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9685
Availability details for
Quantity
  • Availability
  • Add to Cart