COM448628692 Combi-Blocks, Inc.

1-(2-Amino-1-m-tolyl-ethyl)-pyrrolidine-3-carboxylic acid

  • MDL number MFCD07369827
  • Cas Number : 886363-94-6
  • Molecular Weight : 248.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9716

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9716
Availability details for
Quantity
  • Availability
  • Add to Cart