COM448628694 Combi-Blocks, Inc.

1-(2-Aminoethyl)-3-(4-tert-butylthiazol-2-yl)urea

  • MDL number MFCD11501880
  • Cas Number : 1092460-38-2
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9721

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9721
Availability details for
Quantity
  • Availability
  • Add to Cart