COM448628702 Combi-Blocks, Inc.

1-(2-Chlorophenyl)-n1,n1-dimethylethane-1,2-diamine DiHCl

  • MDL number MFCD20921686
  • Cas Number : 1352305-17-9
  • Molecular Weight : 271.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9735

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9735
Availability details for
Quantity
  • Availability
  • Add to Cart