COM448628714 Combi-Blocks, Inc.

1-(3,5-Dimethoxyphenyl)piperazine

  • MDL number MFCD00040764
  • Cas Number : 53557-93-0
  • Molecular Weight : 222.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9758

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9758
Availability details for
Quantity
  • Availability
  • Add to Cart