COM448628716 Combi-Blocks, Inc.

1-(3-Amino-4-(benzyloxy)-2-hydroxyphenyl)ethanone hcl

  • MDL number MFCD21607218
  • Cas Number : 1363404-82-3
  • Molecular Weight : 293.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9763

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9763
Availability details for
Quantity
  • Availability
  • Add to Cart