COM448628729 Combi-Blocks, Inc.

1-(4-Bromophenyl)-1-(4-methoxyphenyl)methylamine

  • MDL number MFCD04115330
  • Cas Number : 886362-84-1
  • Molecular Weight : 292.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9797

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9797
Availability details for
Quantity
  • Availability
  • Add to Cart