COM448628738 Combi-Blocks, Inc.

1-(4-Chlorophenyl)-1-(4-methoxyphenyl)methylamine, HCl

  • MDL number MFCD08436166
  • Cas Number : 856355-54-9
  • Molecular Weight : 284.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9813

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9813
Availability details for
Quantity
  • Availability
  • Add to Cart