COM448628739 Combi-Blocks, Inc.

1-(4-Chlorophenyl)-1,3,8-triazaspiro[4.5]decan-4-one

  • MDL number MFCD18711449
  • Cas Number : 61271-84-9
  • Molecular Weight : 265.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9814

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9814
Availability details for
Quantity
  • Availability
  • Add to Cart