COM448628740 Combi-Blocks, Inc.

1-(4-Chloro-phenyl)-1h-pyrazole

  • MDL number MFCD11845711
  • Cas Number : 25419-86-7
  • Molecular Weight : 178.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9815

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9815
Availability details for
Quantity
  • Availability
  • Add to Cart