COM448628747 Combi-Blocks, Inc.

1-(4-Fluorophenyl)-1,3,8-triazaspiro[4.5]decan-4-one

  • MDL number MFCD18711452
  • Cas Number : 58012-16-1
  • Molecular Weight : 249.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9830

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9830
Availability details for
Quantity
  • Availability
  • Add to Cart