COM448628749 Combi-Blocks, Inc.

1-(4-Methoxyphenyl)-1-phenylethanamine, HCl

  • MDL number MFCD11100898
  • Cas Number : 1189717-77-8
  • Molecular Weight : 263.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9843

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9843
Availability details for
Quantity
  • Availability
  • Add to Cart