COM448628761 Combi-Blocks, Inc.

1-(5-Chloro-2,3-dimethoxy-phenyl)-ethanone

  • MDL number MFCD09800420
  • Cas Number : 117052-19-4
  • Molecular Weight : 214.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9870

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9870
Availability details for
Quantity
  • Availability
  • Add to Cart