COM448628762 Combi-Blocks, Inc.

1-(5-Chloro-2,3-dimethoxyphenyl)ethylamine

  • MDL number MFCD09800421
  • Cas Number : 903581-03-3
  • Molecular Weight : 215.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9871

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9871
Availability details for
Quantity
  • Availability
  • Add to Cart