COM448628766 Combi-Blocks, Inc.

1-(7-(L-Proline-1-yl)-heptyl)-l-proline

  • MDL number MFCD11053582
  • Cas Number : 1820575-06-1
  • Molecular Weight : 326.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9883

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9883
Availability details for
Quantity
  • Availability
  • Add to Cart