COM448628789 Combi-Blocks, Inc.

1,1-Cyclopropanediacetonitrile

  • MDL number MFCD10687162
  • Cas Number : 20778-47-6
  • Molecular Weight : 120.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9927

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9927
Availability details for
Quantity
  • Availability
  • Add to Cart