COM448628790 Combi-Blocks, Inc.

1,1-Dibenzyl-thiourea

  • MDL number MFCD05149016
  • Cas Number : 40398-34-3
  • Molecular Weight : 256.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9928

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9928
Availability details for
Quantity
  • Availability
  • Add to Cart