COM448628792 Combi-Blocks, Inc.

1,1-Dioxo-isothiazolidine-3-carboxylic acid

  • MDL number MFCD07369945
  • Cas Number : 1146957-01-8
  • Molecular Weight : 165.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9930

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9930
Availability details for
Quantity
  • Availability
  • Add to Cart