COM448628813 Combi-Blocks, Inc.

1,3,4,5-Tetrahydro-pyrido[4,3-b]indole-2,8-dicarboxylic acid 2-tert-butyl ester

  • MDL number MFCD04115223
  • Cas Number : 300715-96-2
  • Molecular Weight : 316.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9962

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9962
Availability details for
Quantity
  • Availability
  • Add to Cart