COM448628818 Combi-Blocks, Inc.

1,3-Dihydrospiro[indene-2,4'-piperidine] hcl

  • MDL number MFCD02178983
  • Cas Number : 185525-52-4
  • Molecular Weight : 223.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9976

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9976
Availability details for
Quantity
  • Availability
  • Add to Cart