COM448628819 Combi-Blocks, Inc.

1,3-Dihydroxy-6,6-dimethyl-5,6,7,8-tetrahydroisoquinoline-4-carbonitrile

  • MDL number MFCD20261466
  • Cas Number : 890023-13-9
  • Molecular Weight : 218.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9977

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9977
Availability details for
Quantity
  • Availability
  • Add to Cart