COM448628822 Combi-Blocks, Inc.

1,4-Diaza-spiro[5.5]undecane DiHCl

  • MDL number MFCD06796601
  • Cas Number : 1159822-91-9
  • Molecular Weight : 227.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9985

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OR-9985
Availability details for
Quantity
  • Availability
  • Add to Cart