COM448629141 Combi-Blocks, Inc.

Brimonidine L-Tartrate

  • MDL number MFCD07773072
  • Cas Number : 70359-46-5
  • Molecular Weight : 442.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OS-1329

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OS-1329
Availability details for
Quantity
  • Availability
  • Add to Cart