COM448629277 Combi-Blocks, Inc.

4-Bromo-3-nitropyridine

  • MDL number MFCD04112576
  • Cas Number : 23056-44-2
  • Molecular Weight : 203

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OS-2165

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OS-2165
Availability details for
Quantity
  • Availability
  • Add to Cart