COM448629437 Combi-Blocks, Inc.

3-Amino-2,4-dibromopyridine

  • MDL number MFCD00234046
  • Cas Number : 102249-45-6
  • Molecular Weight : 251.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OS-2382

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OS-2382
Availability details for
Quantity
  • Availability
  • Add to Cart