COM448629884 Combi-Blocks, Inc.

1-Fluoro-2,4-dimethylbenzene

  • MDL number MFCD00060643
  • Cas Number : 452-65-3
  • Molecular Weight : 124.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OS-7811

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OS-7811
Availability details for
Quantity
  • Availability
  • Add to Cart