COM448630023 Combi-Blocks, Inc.

4-Methyl-D-mandelic acid

  • MDL number MFCD00798433
  • Cas Number : 31284-89-6
  • Molecular Weight : 166.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OS-7955

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OS-7955
Availability details for
Quantity
  • Availability
  • Add to Cart