COM448630033 Combi-Blocks, Inc.

Methyl (2R)-2-amino-3-(2-fluorophenyl)propanoate hydrochloride

  • MDL number MFCD24434580
  • Cas Number : 176896-71-2
  • Molecular Weight : 233.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OS-7966

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OS-7966
Availability details for
Quantity
  • Availability
  • Add to Cart