COM448630043 Combi-Blocks, Inc.

2-Oxa-6-azaspiro[3.4]octane oxalic acid

  • MDL number MFCD22123302
  • Cas Number : 1408075-00-2
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OS-7976

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OS-7976
Availability details for
Quantity
  • Availability
  • Add to Cart