COM448630069 Combi-Blocks, Inc.

2-Hydrazinyl-5-methyl-3-nitropyridine

  • MDL number MFCD09027277
  • Cas Number : 21901-25-7
  • Molecular Weight : 168.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-0063

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-0063
Availability details for
Quantity
  • Availability
  • Add to Cart