COM448630176 Combi-Blocks, Inc.

1-(5-Bromo-2-methoxyphenylsulfonyl)piperidine

  • MDL number MFCD02043682
  • Cas Number : 295360-83-7
  • Molecular Weight : 334.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-0199

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-0199
Availability details for
Quantity
  • Availability
  • Add to Cart