COM448630248 Combi-Blocks, Inc.

4-Bromo-N-isopropyl-N-[(4-methoxyphenyl)methyl]benzenesulfonamide

  • MDL number MFCD28397286
  • Cas Number : 1820618-16-3
  • Molecular Weight : 398.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-0405

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-0405
Availability details for
Quantity
  • Availability
  • Add to Cart