COM448630298 Combi-Blocks, Inc.

2-(t-Butyldimethylsilyloxy)-6-bromonaphthalene

  • MDL number MFCD11044858
  • Cas Number : 100751-65-3
  • Molecular Weight : 337.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-0474

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-0474
Availability details for
Quantity
  • Availability
  • Add to Cart