COM448630374 Combi-Blocks, Inc.

1-ethynyl-4-(4-propylcyclohexyl)benzene

  • MDL number MFCD07784127
  • Cas Number : 167858-58-4
  • Molecular Weight : 226.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-0596

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-0596
Availability details for
Quantity
  • Availability
  • Add to Cart