COM448630392 Combi-Blocks, Inc.

1,2-dichloro-4-(methylsulfanyl)benzene

  • MDL number MFCD00028349
  • Cas Number : 17733-23-2
  • Molecular Weight : 193.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-0626

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-0626
Availability details for
Quantity
  • Availability
  • Add to Cart