COM448630464 Combi-Blocks, Inc.

1-(4-t-Butylphenyl)-3-chloropropan-1-one

  • MDL number MFCD11504951
  • Cas Number : 28547-33-3
  • Molecular Weight : 224.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-0733

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-0733
Availability details for
Quantity
  • Availability
  • Add to Cart