COM448630564 Combi-Blocks, Inc.

2-bromo-1-(tert-butoxy)-4-nitrobenzene

  • MDL number MFCD20327985
  • Cas Number : 960309-85-7
  • Molecular Weight : 274.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-0868

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-0868
Availability details for
Quantity
  • Availability
  • Add to Cart