COM448630618 Combi-Blocks, Inc.

Ethyl 2-(4-(4-bromophenylsulfonyl)piperazinoacetate

  • MDL number MFCD12913973
  • Cas Number : 1022353-79-2
  • Molecular Weight : 391.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-0953

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-0953
Availability details for
Quantity
  • Availability
  • Add to Cart