COM448630658 Combi-Blocks, Inc.

5-Bromo-2-(methylsulfonyl)benzotrifluoride

  • MDL number MFCD13195738
  • Cas Number : 300356-32-5
  • Molecular Weight : 303.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-1006

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-1006
Availability details for
Quantity
  • Availability
  • Add to Cart