COM448630689 Combi-Blocks, Inc.

5-Bromo-7-iodo-2,3-dihydro-1-benzofuran

  • MDL number MFCD26904369
  • Cas Number : 1566912-44-4
  • Molecular Weight : 324.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-1140

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-1140
Availability details for
Quantity
  • Availability
  • Add to Cart