COM448630945 Combi-Blocks, Inc.

1-bromo-2,4-dichloro-5-methoxybenzene

  • MDL number MFCD11845926
  • Cas Number : 174913-22-5
  • Molecular Weight : 255.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-1501
Availability details for
Quantity
  • Availability
  • Add to Cart