COM448631079 Combi-Blocks, Inc.

2-(4-Bromo-2-fluorophenyl)ethanol

  • MDL number MFCD19622817
  • Cas Number : 193290-20-9
  • Molecular Weight : 219.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-1687

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-1687
Availability details for
Quantity
  • Availability
  • Add to Cart