COM448631140 Combi-Blocks, Inc.

1-[3-(Benzyloxy)phenyl]-4-methanesulfonylbenzene

  • MDL number MFCD20441885
  • Cas Number : 1345472-18-5
  • Molecular Weight : 338.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-1796

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-1796
Availability details for
Quantity
  • Availability
  • Add to Cart