COM448631238 Combi-Blocks, Inc.

1-bromo-2,5-dichloro-4-fluorobenzene

  • MDL number MFCD11845994
  • Cas Number : 1160573-74-9
  • Molecular Weight : 243.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-1974

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-1974
Availability details for
Quantity
  • Availability
  • Add to Cart