COM448631359 Combi-Blocks, Inc.

tert-butyl 4-(methylcarbamoyl)piperidine-1-carboxylate

  • MDL number MFCD06411659
  • Cas Number : 544696-01-7
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2135

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2135
Availability details for
Quantity
  • Availability
  • Add to Cart