COM448631490 Combi-Blocks, Inc.

1-chloro-3-fluoro-5-iodobenzene

  • MDL number MFCD26792620
  • Cas Number : 1270019-81-2
  • Molecular Weight : 256.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2335

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2335
Availability details for
Quantity
  • Availability
  • Add to Cart