COM448631512 Combi-Blocks, Inc.

N-(1-Phenylethyl)sulfamoyl chloride

  • MDL number MFCD14529134
  • Cas Number : 57053-73-3
  • Molecular Weight : 219.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2381

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2381
Availability details for
Quantity
  • Availability
  • Add to Cart