COM448631560 Combi-Blocks, Inc.

1-(4-Methoxyphenyl)-2-(2-nitrophenyl)ethanone

  • MDL number MFCD23135938
  • Cas Number : 58751-64-7
  • Molecular Weight : 271.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2451

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?OT-2451
Availability details for
Quantity
  • Availability
  • Add to Cart